missing translation for 'onlineSavingsMsg'
Learn More

Butyraldéhyde 98,0+%, TCI America™

Numéro de catalogue. B075125ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B075125ML 25 mL
B0751500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B075125ML Fournisseur TCI America Code fournisseur B075125ML

Identifiants chimiques

CAS 123-72-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00007023
InChI Key ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
Synonym butyraldehyde, n-butyraldehyde, butyral, 1-butanal, butyric aldehyde, butanaldehyde, butaldehyde, butalyde, butal, n-butanal
PubChem CID 261
ChEBI CHEBI:15743
IUPAC Name butanal
SMILES CCCC=O

Spécifications

Color Jaune
Boiling Point 75°C
Quantity 25 mL
UN Number 1129
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquide
Chemical Name or Material Butyraldehyde
EINECSNumber (2)-0494
RTECSNumber ES2275000
TSCA Yes