missing translation for 'onlineSavingsMsg'
Learn More

Butylparaben Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015947
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015947 10 g
111015946 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015947 Supplier Merck Emd Millipore Supplier No. PHR102210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 94-26-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C11H14O3
MDL Number MFCD00016478
Synonym 4-Hydroxybenzoic acid butyl ester
Type Certified Reference Material
Quantity 10 g
Formula Weight 194.23
Grade Certified Reference Material