missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butylmalonic Acid 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 103°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 160.17 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Butylmalonic Acid |
Sécurité et manipulation
| TSCA | No |