missing translation for 'onlineSavingsMsg'
Learn More

Butylboronic Acid (contains varying amounts of Anhydride) [for Esterification], TCI America™

Catalog No. B05291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B05291G Supplier TCI America Supplier No. B05291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4426-47-5
Molecular Formula C4H11BO2
Molecular Weight (g/mol) 101.94
MDL Number MFCD00002106
InChI Key QPKFVRWIISEVCW-UHFFFAOYSA-N
Synonym n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane
PubChem CID 20479
IUPAC Name butylboronic acid
SMILES CCCCB(O)O

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 101.94
Physical Form Crystalline Powder
Chemical Name or Material Butylboronic Acid (contains varying amounts of Anhydride) [for Esterification]