missing translation for 'onlineSavingsMsg'
Learn More

Butylated hydroxyanisole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012183
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012183 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012183 Supplier Merck Emd Millipore Supplier No. PHR13041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 25013-16-5
Ignition Point 599°F
Vapor Density 6.2 (vs. air)
Flash Point 113°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (CH3)3 CC6H3(OCH3)OH
MDL Number MFCD01779059
Melting Point 58°C to 60°C
Synonym 2(3)-t-Butyl-4-hydroxyanisole; 2(3)-t-Butylhydroquinone monomethyl ether; BHA
Type Certified Reference Material
Quantity 1 g
Formula Weight 180.24
Grade Certified Reference Material
Show More Show Less