missing translation for 'onlineSavingsMsg'
Learn More

Butylamine Hydrochloride 98.0+%, TCI America™

Catalog No. B0710500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0710500G 500 g
B071025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0710500G Supplier TCI America Supplier No. B0710500G
Only null left

Chemical Identifiers

CAS 3858-78-4
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.597
MDL Number MFCD00043791
InChI Key ICXXXLGATNSZAV-UHFFFAOYSA-N
Synonym 1-Aminobutane Hydrochloride, Butylammonium Chloride
PubChem CID 6432400
IUPAC Name butan-1-amine;hydrochloride
SMILES CCCCN.Cl

Specifications

Melting Point 214°C
Color White
Quantity 500 g
Formula Weight 109.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Butylamine Hydrochloride
EINECSNumber (2)-0132
RTECSNumber EO5199990
TSCA Yes