missing translation for 'onlineSavingsMsg'
Learn More

Butyl stearate, tech.

Numéro de catalogue. p-7050260
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA3633836 500 g
AA36338A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA3633836 Fournisseur Thermo Scientific Chemicals Code fournisseur 03633836
Il en reste null

CAS: 123-95-5 | C22H44O2 | 340.59 g/mol

Butyl stearate is used as finishing agents, lubricants and lubricant additives. It is also used as a plasticizer, food packaging material and as a dye solvent. It acts as a reactant and involved in the preparation of octadecanoic acid methyl ester by reacting with methanol. It finds application as a binder in cosmetics like soaps, shampoos and shaving creams, skin conditioners and surfactants for cosmetic formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 123-95-5
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.59
MDL Number MFCD00026669
InChI Key ULBTUVJTXULMLP-UHFFFAOYSA-N
Synonym butyl stearate, n-butyl stearate, octadecanoic acid, butyl ester, kesscoflex bs, n-butyl octadecanoate, stearic acid, butyl ester, butyl octadecylate, kessco bsc, stearic acid n-butyl ester, polycizer 332
PubChem CID 31278
ChEBI CHEBI:85983
IUPAC Name butyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCCC

Spécifications

Melting Point 17°C to 22°C
Density 0.861g/mL
Boiling Point 343°C
Flash Point 160° (320°F)
Quantity 500 g
Beilstein 1792866
Merck Index 14,1589
Formula Weight 340.59
Grade Technical
Physical Form Liquid
Chemical Name or Material Butyl Stearate
Afficher plus de résultats Afficher moins de résultats
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a
RTECSNumber WI2900000
TSCA TSCA

RUO – Research Use Only