missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyl Palmitate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 170°C |
| Quantity | 25 mL |
| Formula Weight | 312.54 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Butyl Palmitate |
Safety and Handling
| EINECSNumber | (2)-0798 |
| TSCA | Yes |