missing translation for 'onlineSavingsMsg'
Learn More

Butyl p-Toluenesulfonate 97.0+%, TCI America™

Catalog No. T072725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T072725ML 25 mL
T0727500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T072725ML Supplier TCI America Supplier No. T072725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 778-28-9
Molecular Formula C11H16O3S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00027203
InChI Key QYJXDIUNDMRLAO-UHFFFAOYSA-N
Synonym butyl p-toluenesulfonate, butyl tosylate, n-butyl p-toluenesulfonate, butyl p-methylbenzenesulfonate, butyl-p-toluenesulfonate, p-toluenesulfonic acid, butyl ester, benzenesulfonic acid, 4-methyl-, butyl ester, butyl toluene-4-sulphonate, p-toluenesulfonic acid n-butyl ester, p-toluene sulfonic acid, butyl ester
PubChem CID 13066
IUPAC Name butyl 4-methylbenzene-1-sulfonate
SMILES CCCCOS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 mL
Formula Weight 228.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl p-Toluenesulfonate
EINECSNumber (3)-1911
RTECSNumber XT6400000
TSCA Yes