missing translation for 'onlineSavingsMsg'
Learn More

Butyl Alcohol, Reagent, ACS, 99.4%, Spectrum™ Chemical

B1185, 71-36-3, C4H10O

Supplier:  Spectrum Chemical Mfg Cor B1185500MLGL

 View more versions of this product

Catalog No. 18601154


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Butyl Alcohol, Reagent, ACS is more commonly known as butanol. It is also widely used as a solvent in textile and chemical fabrication processes. It can be used as a paint thinner and base for perfumes. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Specifications

Specifications

Butyl Alcohol
100%
500 mL
0.005%
C4H10O
0.1%
CCCCO
74.12
Reagent
71-36-3
117°C
Amber Glass Bottle
0.0008 meq/g
UN1120
LRHPLDYGYMQRHN-UHFFFAOYSA-N
butan-1-ol
99.4%

Chemical Identifiers

71-36-3
74.12
butan-1-ol
C4H10O
LRHPLDYGYMQRHN-UHFFFAOYSA-N
CCCCO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions