missing translation for 'onlineSavingsMsg'
Learn More

Butyl Alcohol, NF, 99.5%, Spectrum™ Chemical

Catalog No. 18601145
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601145 500 mL Amber Glass Bottle
18601147 1 L Amber Glass Bottle
18601146 6 x 500 mL Amber Glass Bottle
18601148 4 L Amber Glass Bottle
18601149 6 x 1 L Amber Glass Bottle
18601150 4 x 4 L Amber Glass Bottle
18601152 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18601145 Supplier Spectrum Chemical Mfg Cor Supplier No. B1197500MLGL
Only null left

B1197, 71-36-3, C4H10O

Spectrum™ Chemical Butyl Alcohol, NF is primarily a solvent and vehicle in many pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Max % 100%
Quantity 500 mL
Packaging Amber Glass Bottle
UN Number UN1120
Boiling Point 117°C
Percent Purity 99.5%
Grade NF
Chemical Name or Material Butyl Alcohol