missing translation for 'onlineSavingsMsg'
Learn More

Butyl acrylate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020662
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020662 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020662 Supplier Merck Emd Millipore Supplier No. 069581MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Environmental

Specifications

CAS 141-32-2
Ignition Point 559°F
Solution Contains ˜0.0015% hydroquinone monomethyl ether as stabilizer
Vapor Pressure 3.3 mmHg (20°C)
Vapor Density >1 (vs. air)
Density 0.899 g/mL (at 20°C)
Flash Point 36°C
Linear Formula CH2=CHCOO(CH2)3 CH3
MDL Number MFCD00009446
Boiling Point 145°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 128.17
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less