missing translation for 'onlineSavingsMsg'
Learn More

Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7%, Solstice

Catalog No. 6001011
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001011 1 L Glass Bottle
6001012 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001011 Supplier Solstice Supplier No. 2706871L

CHROMASOLV™ Plus, for HPLC, 99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 123-86-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009445
InChI Key DKPFZGUDAPQIHT-UHFFFAOYSA-N
Synonym n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle
PubChem CID 31272
ChEBI CHEBI:31328
IUPAC Name butyl acetate
SMILES CCCCOC(C)=O

Specifications

Absorbance max. 0.01 at 400 nm, max. 0.01 at 320 nm, max. 0.02 at 300 nm, max. 0.04 at 275 nm, max. 0.20 at 260 nm
CAS Max % 100.0000%
Melting Point -78°C (literature)
Density 0.88 g/mL at 25 °C(lit.)
Boiling Point 126.2°C
Flash Point 27°C
Packaging Glass Bottle
Linear Formula CH3COO(CH2)3CH3
Quantity 1 L
UN Number UN1123
Beilstein 1741921
Formula Weight 116.16
Percent Purity >99.65%
Grade HPLC
Chemical Name or Material Butyl acetate, CHROMASOLV™ Plus, for HPLC, 99.7%
Show More Show Less
ShelfLife 900 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Butyl acetates
EINECSNumber 204-658-1
Recommended Storage Room Temp