missing translation for 'onlineSavingsMsg'
Learn More

Butyl 4-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. H0210500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0210500G 500 g
H021025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0210500G Supplier TCI America Supplier No. H0210500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-26-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00016478
InChI Key QFOHBWFCKVYLES-UHFFFAOYSA-N
Synonym butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl
PubChem CID 7184
IUPAC Name butyl 4-hydroxybenzoate
SMILES CCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 70°C
Color White
Boiling Point 156°C
Quantity 500 g
Formula Weight 194.23
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Butyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
RTECSNumber DH1980000
TSCA Yes