missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Burkholderia cepacia Sel Agar (USP 60) 500g

Catalog No. R452642
Change view
Click to view available options
500g container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R452642 500g container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R452642 Supplier Thermo Scientific™ Supplier No. R452642

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Compliance to the USP 60 regulations for B. cepacia complex made simple

Burkholderia cepacia medium (USP 60) is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in products, especially those for inhalation use of non-sterile acqueous preparations for oral, oromucosal, cutaneous or nasal use.

Confidently detect the presence of B. cepacia in nonsterile medicine with a complete workflow for USP 60 requirements which satisfies all regulations.

  • Compliant formulation according to USP 60
  • Quality tested to meet USP 60 specifically for Pharma customers
  • Part of a complete workflow which include DCM and QCO

Specifications

Description Burkholderia cepacia Selective agar (USP 60)
Format 500g Pot
Product Type Agar
For Use With (Application) Pharmaceutical testing of non-sterile products used for nasal inhalation
Quantity 500 g
Regulatory Status Lab use only