missing translation for 'onlineSavingsMsg'
Learn More

Bupropion Related Compound F Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015693
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015693 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015693 Supplier Merck Emd Millipore Supplier No. PHR175530MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 857233-13-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C9H9 ClO2
Synonym 1-(3-Chlorophenyl)-1-hydroxy-2-propanone
Type Certified Reference Material
Quantity 30 mg
Formula Weight 184.62
Grade Certified Reference Material