missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avertissement:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Identifiants chimiques
| Halogen Lamp |
Spécifications
| 2 Pin (G4) | |
| 100 hr. | |
| 20 W | |
| 0.35 in. | |
| CBAR6 | |
| 1.21 in. | |
| 3.07 cm | |
| 480 lm | |
| Halogen Lamp |
| Halogen | |
| 6 V | |
| Clear | |
| 0.8 cm | |
| Microscope, diagnostic instrument | |
| 0.76 in. | |
| 1.93 cm | |
| T3 |