missing translation for 'onlineSavingsMsg'
Learn More

Bulbtronics™ Halogen Lamps

Numéro de catalogue. 14395366
Encompass
Change view
Click to view available options
Length (English):
1.21 in.
1.73 in.
3.54 in.
Base Style:
2 Pin (G4)
2 Pin (G6.35)
SFA7.5-2, SFA9-2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Length (English) Base Style
14395366 3.54 in. SFA7.5-2, SFA9-2
14395367 3.54 in. 2 Pin (G4)
14395414 1.21 in. 2 Pin (G4)
14395427 1.73 in. 2 Pin (G6.35)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Numéro de catalogue. 14395366 Fournisseur Bulbtronics™ Code fournisseur 256
Il en reste null

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Spécifications

Base Style SFA7.5-2, SFA9-2
Light Source Mercury Arc
Lighting Life 200 hr.
Wattage 100 W
Color Clear
Diameter (English) 0.39 in.
Diameter (Metric) 0.99 cm
For Use With (Equipment) Microscope
Length (English) 3.54 in.
Length (Metric) 9 cm
Lumen 2200 lm
Product Type Halogen Lamp
Afficher plus de résultats Afficher moins de résultats