missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 12.45 at 25°C, Certified, LabChem™

Supplier:  LabChem LC125624

 View more versions of this product

Catalog No. LC125624


Only null left

Description

Description

Specifications

Specifications

Buffer Solution pH 12.45 at 25°C
99.49,0.35,0.16
Phosphine; Phosphorus oxides; Phosgene; Thermal decomposition generates: Corrosive vapors
12.45 ±0.05
4 L
Na2HPO4
1g/mL
Soluble in water
Certified
Colorless
7558-79-4 , 1310-73-2
1g/mL
Liquid
Passes Test
Poly Bottle
Traceable to NIST
141.96
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions