missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 12.45 at 25°C, Certified, LabChem™
Supplier: LabChem LC125621
Specifications
| Buffer Solution pH 12.45 at 25°C | |
| 99.49,0.35,0.16 | |
| Phosphine; Phosphorus oxides; Phosgene; Thermal decomposition generates: Corrosive vapors | |
| 12.45 ±0.05 | |
| 500 mL | |
| Na2HPO4 | |
| 1g/mL | |
| Soluble in water | |
| Certified |