missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 12.0 at 25°C, LabChem™
Supplier: LabChem LC125602
Specifications
| Buffer Solution pH 12.0 at 25°C | |
| 99.58,0.35,0.07 | |
| Phosphine; Phosphorus oxides; Phosgene; Thermal decomposition generates: Corrosive vapors | |
| 12.00 ±0.05 | |
| 1 L | |
| Na2HPO4 | |
| 1g/mL | |
| Soluble in water | |
| Certified |