missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 1.68 at 25°C, Certified, LabChem™

$65.14 - $258.04

Specifications

Chemical Name or Material Buffer Solution pH 1.68 at 25°C
CAS Max % 98.55,1.26,0.19
Color Colorless
CAS 6153-56-6 , 1310-58-3
Decomposition Information Potassium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC122101
View Documents
LabChem
LC122101
500 mL
Each for $65.14
Only null left
 
LC122102
View Documents
LabChem
LC122102
1 L
Each for $104.82
Only null left
 
LC122104
View Documents
LabChem
LC122104
4 L
Each for $258.04
Only null left
 
Description

Description

Specifications

Specifications

Buffer Solution pH 1.68 at 25°C
Colorless
Potassium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
1.68 ±0.01
Passes Test
Poly Bottle
Traceable to NIST
90.04
98.55,1.26,0.19
6153-56-6 , 1310-58-3
1g/mL
Liquid
HO2CCO2H
1g/mL
Soluble in water
Certified
Videos
SDS
Documents

Documents