missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 1.68 at 25°C, Certified, LabChem™
Supplier: LabChem LC122102
Specifications
| Buffer Solution pH 1.68 at 25°C | |
| 98.55,1.26,0.19 | |
| Potassium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors | |
| 1.68 ±0.01 | |
| 1 L | |
| HO2CCO2H | |
| 1g/mL | |
| Soluble in water | |
| Certified |