missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 1.68 at 25°C, Certified, LabChem™

Supplier:  LabChem LC122101

 View more versions of this product

Catalog No. LC122101


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Buffer Solution pH 1.68 at 25°C
98.55,1.26,0.19
Potassium oxide; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
1.68 ±0.01
500 mL
HO2CCO2H
1g/mL
Soluble in water
Certified
Colorless
6153-56-6 , 1310-58-3
1g/mL
Liquid
Passes Test
Poly Bottle
Traceable to NIST
90.04
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions