missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution (Acetate), pH 4.00 (Certified), Fisher Chemical

Catalog No. SB851
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB851 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB851 Supplier Thermo Fisher Scientific Supplier No. BPSB851
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with chlorine titrators.

Specifications

Chemical Name or Material Buffer Solution (Acetate), pH 4.01
CAS 64-19-7, 127-09-3, 50-00-0, 67-56-1, 7732-18-5
Color Undesignated
pH 4.00
Physical Form Liquid
Quantity 1 L
Identification Pass Test
Packaging Poly Bottle
MDL Number MFCD00137248
Traceability to NIST Traceable to NIST.
Synonym Chlorine Titrimeter Reagent
Grade Certified
Show More Show Less