missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 9.18 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 159525
Change view
Click to view available options
Package: Cubitainer; Size: 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
159525 Package: Cubitainer; Size: 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 159525 Supplier Ricca Chemical Company Supplier No. 15952.5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 9.18± 0.01 at 25°C
CAS 1303-96-4, 7732-18-5
CAS Min % 98.0, 0.37
CAS Max % 100.0, 0.39
pH 9.18
Quantity 10 L
Packaging Cubitainer
Sterility Non-sterile
Grade Reference Standard