missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 7.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 361018145
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361018145 1 L Natural Poly Bottle
361018144 500 mL Natural Poly Bottle
15501CT 4 L Cubitainer
15501 4 L Natural Poly Bottle
155025 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361018145 Supplier Ricca Chemical Company Supplier No. 155032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 7.00 ± 0.01 at 25 deg C
CAS 7558-79-4, 7778-77-0, 7732-18-5
CAS Min % 97.0, 0.87, 0.48
CAS Max % 100.0, 0.91, 0.5
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 7
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Packaging Natural Poly Bottle
Show More Show Less