missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 6.86 ± 0.01 at 25°C, Ricca Chemical
$72.57 - $442.66
Specifications
| CAS | 7558-79-4,7778-77-0,7732-18-5 |
|---|---|
| CAS Min % | 97.0,0.34,0.33 |
| CAS Max % | 100.0,0.36,0.34 |
| Color | Colorless |
| Concentration | 1X |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
36-101-8142
|
Ricca Chemical Company
154016 |
500 mL | Natural Poly Bottle |
Each for $72.57
|
|
||||
|
36-101-8143
|
Ricca Chemical Company
154032 |
1 L | Natural Poly Bottle |
Each for $112.50
|
|
||||
|
R1540-1
|
Ricca Chemical Company
15401 |
4 L | Natural Poly Bottle |
Each for $220.31
|
|
||||
|
1540-25
|
Ricca Chemical Company
154025 |
10 L | Cubitainer |
Each for $358.81
|
|
||||
|
1540-5
|
Ricca Chemical Company
15405 |
20 L | Cubitainer |
Each for $442.66
|
|
||||
Specifications
| 7558-79-4,7778-77-0,7732-18-5 | |
| 100.0,0.36,0.34 | |
| 1X | |
| pH Meter and Electrode | |
| Liquid |
| 97.0,0.34,0.33 | |
| Colorless | |
| Calibration | |
| 6.86 | |
| pH Buffer |