missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 6.86 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 361018142
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361018142 500 mL Natural Poly Bottle
361018143 1 L Natural Poly Bottle
R15401 4 L Natural Poly Bottle
154025 10 L Cubitainer
15405 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 361018142 Supplier Ricca Chemical Company Supplier No. 154016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 6.86± 0.01 at 25°C
CAS 7558-79-4, 7778-77-0, 7732-18-5
CAS Min % 97.0, 0.34, 0.33
Color Colorless
CAS Max % 100.0, 0.36, 0.34
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 6.86
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Reference Standard
Show More Show Less