missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 6.86 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company 154032
Specifications
| Buffer, Reference Standard | |
| 7558-79-4,7778-77-0,7732-18-5 | |
| Colorless | |
| 1X | |
| pH Meter and Electrode | |
| Liquid | |
| pH Buffer | |
| Non-sterile |
| pH 6.86± 0.01 at 25°C | |
| 97.0,0.34,0.33 | |
| 100.0,0.36,0.34 | |
| Calibration | |
| 6.86 | |
| 1 L | |
| Natural Poly Bottle | |
| Reference Standard |