missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 5.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 361018138
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361018138 500 mL Natural Poly Bottle
R150500020 20 x 20 mL Pack
361018139 1 L Natural Poly Bottle
15051 4 L Natural Poly Bottle
150525 10 L Cubitainer
15055 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 361018138 Supplier Ricca Chemical Company Supplier No. 150516
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 5.00 ± 0.01 at 25°C
CAS 877-24-7, 7732-18-5
Color Colorless
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 5
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Natural Poly Bottle
Grade Not Available
Show More Show Less