missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 4.63 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 150416

Catalog No. 36-101-8137


Only null left

Description

Description

Specifications

Specifications

Buffer, Reference Standard
877-24-7 , 1310-73-2 , 7732-18-5
Colorless
1X
pH Meter and Electrode
Liquid
pH Buffer
pH 4.63± 0.01 at 25°C
97.0,0.99,0.07
100.0,1.03,0.07
Calibration
4.63
500 mL
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions