missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 2.00 ± 0.01 at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company R14930001A

 View more versions of this product

Catalog No. 1493-32


Only null left

Description

Description

Specifications

Specifications

Buffer, Reference Standard
0°C
100°C
1X
pH Meter and Electrode
Liquid
pH Buffer
Miscible
pH 2.00± 0.01 at 25°C
7447-40-7 , 7732-18-5
Colorless
Calibration
2
1 L
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions