missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 12.00 ± 0.01 at 25°C, Ricca Chemical
$39.75 - $369.66
Specifications
| Chemical Name or Material | Buffer, Reference Standard |
|---|---|
| Name Note | pH 12.00± 0.01 at 25°C |
| CAS | 7447-40-7,1310-73-2,7732-18-5 |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R161500020P
|
Ricca Chemical Company
R161500020PK |
20 x 20 mL | Pack |
Each for $109.63
|
|
||||
|
1615-16
|
Ricca Chemical Company
161516 |
500 mL | Clear PET-G Bottle |
Each for $50.47
|
|
||||
|
161532
|
Ricca Chemical Company
161532 |
1 L | Natural Poly Bottle |
Each for $39.75
|
|
||||
|
16151
|
Ricca Chemical Company
16151 |
4 L | Natural Poly Bottle |
Each for $197.30
|
|
||||
|
16155
|
Ricca Chemical Company
RI16155 |
20 L | Cubitainer |
Each for $369.66
|
|
||||
Specifications
| Buffer, Reference Standard | |
| 7447-40-7,1310-73-2,7732-18-5 | |
| 100°C | |
| 100.0,0.38,0.08 | |
| 1X | |
| pH Meter and Electrode | |
| Liquid | |
| Non-sterile | |
| Reference Standard |
| pH 12.00± 0.01 at 25°C | |
| 0°C | |
| 98.0,0.36,0.08 | |
| Colorless | |
| Calibration | |
| 12 | |
| pH Buffer | |
| Miscible |