missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 12.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 161516
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
20 L
Packaging:
Clear PET-G Bottle
Cubitainer
Natural Poly Bottle
Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
161516 500 mL Clear PET-G Bottle
R161500020P 20 x 20 mL Pack
161532 1 L Natural Poly Bottle
16151 4 L Natural Poly Bottle
16155 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 161516 Supplier Ricca Chemical Company Supplier No. 161516
Edge

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 12.00± 0.01 at 25°C
Melting Point 0°C
CAS 7447-40-7, 1310-73-2, 7732-18-5
Boiling Point 100°C
CAS Min % 98.0, 0.36, 0.08
Color Colorless
CAS Max % 100.0, 0.38, 0.08
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 12
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer
Packaging Clear PET-G Bottle
Sterility Non-sterile
Solubility Information Miscible
Grade Reference Standard
Show More Show Less