missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer, Reference Standard, pH 12.00 ± 0.01 at 25°C, Ricca Chemical
Supplier: Ricca Chemical Company RI16155
Specifications
| Buffer, Reference Standard | |
| 0°C | |
| 100°C | |
| 100.0,0.38,0.08 | |
| 1X | |
| pH Meter and Electrode | |
| Liquid | |
| pH Buffer | |
| Non-sterile | |
| Reference Standard |
| pH 12.00± 0.01 at 25°C | |
| 7447-40-7,1310-73-2,7732-18-5 | |
| 98.0,0.36,0.08 | |
| Colorless | |
| Calibration | |
| 12 | |
| 20 L | |
| Cubitainer | |
| Miscible |