missing translation for 'onlineSavingsMsg'
Learn More

Buffer, for Fluoride, Citrate - Acetate, Ricca Chemical

Catalog No. 147816
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
147816 500 mL
14781 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 147816 Supplier Ricca Chemical Company Supplier No. 147816

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer
CAS 6132-04-3, 64-19-7, 7647-14-5, 1310-73-2, 6132-04-3
CAS Min % 85.0, 4.52, 4.39, 3.86, 0.02
Color Colorless
CAS Max % 89.0, 4.71, 4.57, 4.02, 0.02
Physical Form Liquid
Quantity 500 mL
Molecular Formula C6H9Na3O9
Packaging Natural Poly Bottle
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Molecular Weight (g/mol) 294.10
Show More Show Less