missing translation for 'onlineSavingsMsg'
Learn More

Buffer Concentrate, pH 7.00 (Certified), Fisher Chemical™

Catalog No. SB109500
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB109500 500 mL
SB1091 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB109500 Supplier Thermo Fisher Scientific Supplier No. FLSB109500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer Concentrate, pH 7.00
CAS 64-02-8, 7732-18-5, 1310-58-3, 1310-73-2, 7664-38-2
Physical Form Liquid
Quantity 500 mL
Identification Pass Test
Molecular Formula C10H12N2Na4O8
MDL Number MFCD00146206
Packaging Poly Bottle
Traceability to NIST Traceable to NIST.
InChI Key UEUXEKPTXMALOB-UHFFFAOYSA-J
Molecular Weight (g/mol) 380.17
Grade Certified
Show More Show Less