missing translation for 'onlineSavingsMsg'
Learn More

Buffer Concentrate, pH 4.00 (Certified), Fisher Chemical™

Catalog No. SB991
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB991 1 L
SB99500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SB991 Supplier Thermo Fisher Scientific Supplier No. BPSB991
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer Concentrate, pH 4.00
Melting Point 0°C
CAS 877-24-7, 50-00-0, 67-56-1, 7732-18-5
Boiling Point 100°C
Color Undesignated
Density 1.0g/cm³
pH 4.0
Physical Form Liquid
Quantity 1 L
Identification Pass Test
Packaging Poly Bottle
MDL Number MFCD00013070
Traceability to NIST Traceable to NIST.
Vapor Pressure 14mmHg at 20°C
Grade Certified
Show More Show Less