missing translation for 'onlineSavingsMsg'
Learn More

Budesonide 98.0+%, TCI America™

Catalog No. B3909200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3909200MG 200 mg
B39091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3909200MG Supplier TCI America Supplier No. B3909200MG

Chemical Identifiers

CAS 51333-22-3
Molecular Formula C25H34O6
Molecular Weight (g/mol) 430.54
MDL Number MFCD00083259,MFCD00138154
InChI Key VOVIALXJUBGFJZ-VXKMTNQYSA-N
Synonym (+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione
PubChem CID 73759663
IUPAC Name (1S,2S,4R,6R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-16-one
SMILES CCC[C@@H]1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 430.54
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Budesonide
RTECSNumber TU3723000
TSCA No