missing translation for 'onlineSavingsMsg'
Learn More

Budesonide Related Compound L Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013547
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013547 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013547 Supplier Merck Emd Millipore Supplier No. PHR182350MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 216453-74-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C25H32O6
Synonym 16 alpha,17-[Butylidenebis(oxy)]-21-hydroxypregna-1,4-diene-3,11,20-trione; 16 alpha,17-[butylidenebis(oxy)]-21-hydroxypregna-1,4-diene-3,11,20-trione; 11-Ketobudesonide
Type Certified Reference Material
Quantity 50 mg
Formula Weight 428.52
Grade Certified Reference Material