Chemglass Inc Vapor Tube, Steam Duct, 160mm, 24/40 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 285mm Approx OAL
Numéro de catalogue. CG131721
Vapor Tube, Steam Duct, 160mm, 24/40 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 285mm Approx OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the 4 listed sizes. NOT SUPPLIED WITH COMBI-CLIP.Dimension A:CG-1317-21: 160mm
Numéro de catalogue.
CG131721
Fournisseur
Chemglass IncCode fournisseur
CG131721
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Vapor Tube, Steam Duct, 160mm, 24/40 Joint, Compatible with Buchi Rotary Evaporators, Combi-Clip Not Included, 285mm Approx OAL. Replacement Vapor tubes compatible with Buchi rotary evaporators. Thread is made for use with standard Combi-Clip. Available in the 4 listed sizes. NOT SUPPLIED WITH COMBI-CLIP.Dimension A:CG-1317-21: 160mm