missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrodes: Handle Only
Description
Comprend
8cm long array handle
s
- Reusable
- 45-0167 handle designed for 45-0120 10 mm needle tips
- Used for larger muscle masses
- 45-0168 handle designed for 45-0121 5 mm needle tips
- Recommended for smaller muscle masses
Spécifications
Spécifications
| For Use With (Application) | In-Vivo Drug or Gene Delivery, Muscle Gene Therapy |
| For Use With (Equipment) | 2-needle Array (45-0167), ECM830, Gemini X2 and ECM2001+ Generators |
| Material | Delrin |
| Product Line | BTX Electroporation |
| Product Type | Electroporation accessory |
| Voltage | 500 V |
| Description | 2-Needle Array Handle, 10 mm |
| Includes | Electrode handle only (requires 45-0167) |
| Length (English) | 3.2 in. |
| Length (Metric) | 80 mm |
| Afficher plus de résultats |