missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Two-Needle Array™ Electrode Kit

Two-Needle Array™ Electrode Kit designed for in vivo drug or gene delivery.

$1,510.51 - $1,655.94

Specifications

For Use With (Application) In-Vivo Drug or Gene Delivery, Muscle Gene Therapy
Material Stainless Steel, Delrin
Product Line BTX Electroporation
Voltage 500 V
Length (English) Electrode 0.78 in.
View More Specs

Includes:
  • Handle
  • Six two-needle arrays

Products 2
Catalog Number Mfr. No. Size (Metric) Gap Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Gap Price Quantity & Availability  
BTX450167
View Documents Promotion Details
BTX™
45 0167
10 mm
Each for $1,655.94
Only null left
 
BTX450168
View Documents Promotion Details
BTX™
45 0168
5 mm
Each for $1,510.51
Only null left
 
Description

Description

  • Consists of reusable array handle and sterile, disposable two-needle array
  • Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
  • Compatible with ECM™ 830, ECM 2001, ECM 630
  • Supplied in convenient kit with Delrin handle and 20 mm stainless-steel needle array (pack of 6), or order components separately (match handle size to needle array size)

Two Sizes

  • 5 mm array and handle recommended for small muscle masses such as mouse tibialis
  • 10 mm size suitable for larger muscle masses such as rat gastronomes
Specifications

Specifications

In-Vivo Drug or Gene Delivery, Muscle Gene Therapy
BTX Electroporation
0.78 in.
10 μsec. to 99 msec. in PBS
Non-sterile
Square Wave
Stainless Steel, Delrin
500 V
20 mm
5°C to 40°C
Banana Cables
Videos
SDS
Documents

Documents

Intended for Research use only. Not for use in diagnostic or clinical procedures.