missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Two-Needle Array™ Electrode Kit
Two-Needle Array™ Electrode Kit designed for in vivo drug or gene delivery.
Supplier: BTX™ 45 0167
Includes:
- Handle
- Six two-needle arrays
Description
- Consists of reusable array handle and sterile, disposable two-needle array
- Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
- Compatible with ECM™ 830, ECM 2001, ECM 630
- Supplied in convenient kit with Delrin handle and 20 mm stainless-steel needle array (pack of 6), or order components separately (match handle size to needle array size)
Two Sizes
- 5 mm array and handle recommended for small muscle masses such as mouse tibialis
- 10 mm size suitable for larger muscle masses such as rat gastronomes
Chemical Identifiers
| In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
| BTX Electroporation | |
| 0.78 in. | |
| 10 μsec. to 99 msec. in PBS | |
| Non-sterile | |
| Square Wave |
| Stainless Steel, Delrin | |
| 500 V | |
| 20 mm | |
| 5°C to 40°C | |
| Banana Cables |
Specifications
| In-Vivo Drug or Gene Delivery, Muscle Gene Therapy | |
| Stainless Steel, Delrin | |
| Electroporation accessories | |
| 2-Needle Array Kit, 10 mm, Pkg. of 6, with Handle | |
| 6 x 10 mm 2-needle Arrays, 1 x Reusable Handle | |
| 20 mm | |
| 5°C to 40°C | |
| 10 mm | |
| Non-sterile | |
| Square Wave |
| ECM 830, ECM 2001+, ECM 2001, Gemini X2 Generator | |
| BTX Electroporation | |
| 500 V | |
| Twin needle | |
| 0.78 in. | |
| 10 μsec. to 99 msec. in PBS | |
| 0.39 in. | |
| Needle array | |
| Banana Cables |
Intended for Research use only. Not for use in diagnostic or clinical procedures.