missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Two-Needle Array™ Electrode Kit

Catalog No. BTX450168
Change view
Click to view available options
Size (Metric) Gap:
5 mm
10 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Gap
BTX450168 5 mm
BTX450167 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX450168 Supplier BTX™ Supplier No. 45 0168

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Handle
  • Six two-needle arrays

Two-Needle Array™ Electrode Kit designed for in vivo drug or gene delivery.

  • Consists of reusable array handle and sterile, disposable two-needle array
  • Enables simple, inexpensive, safe direct injection of muscle tissue for plasmid DNA delivery
  • Compatible with ECM™ 830, ECM 2001, ECM 630
  • Supplied in convenient kit with Delrin handle and 20 mm stainless-steel needle array (pack of 6), or order components separately (match handle size to needle array size)

Two Sizes

  • 5 mm array and handle recommended for small muscle masses such as mouse tibialis
  • 10 mm size suitable for larger muscle masses such as rat gastronomes

Specifications

For Use With (Application) In-Vivo Drug or Gene Delivery, Muscle Gene Therapy
For Use With (Equipment) ECM 830, Gemini X2, ECM 2001, ECM 2001+Generator
Material Stainless Steel, Delrin
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 500 V
Description 2-Needle Array Kit, 5 mm, Pkg. of 6, with Handle
Electrode Shape Straight/Needle
Includes 6 x 5 mm 2-needle Arrays, 1 x Reusable Handle
Length (English) Electrode 0.78 in.
Length (Metric) Electrode 20 mm
Pulse Length Range 10 μsec. to 99 msec. in PBS
Operating Temperature (Metric) 5°C to 40°C
Size (English) Gap 0.19 in.
Size (Metric) Gap 5 mm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.