missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Genetrodes™ Electrodes

Catalog No. BTX516
Change view
Click to view available options
Electrode Shape:
Bent L-shaped
Straight
Length (Metric) Electrode:
1 mm
3 mm
5 mm
10 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrode Shape Length (Metric) Electrode
BTX516 Bent L-shaped 1 mm
BTX514 Bent L-shaped 3 mm
BTX512 Bent L-shaped 5 mm
BTX510 Straight 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BTX516 Supplier BTX™ Supplier No. 45 0117

Needle-style reusable Genetrodes™ Electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems.

  • Needle-style reusable electrodes compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Five models available
  • Models 508 and 510 are straight with gold tips designed for general in vivo applications
  • Models 512, 514, and 516 are L-shaped electrodes with gold tips designed for in vivo and in ovo electroporation

Order Info

Genetrode holder with shaft available separately (Cat. No. BTX515).

Specifications

For Use With (Application) In-Vivo Gene Delivery, ex-Vivo Gene Delivery, in-Ovo Gene Delivery
For Use With (Equipment) ECM 830, ECM 2001+, ECM 2001, and Gemini X2
Material Gold
Product Line BTX Electroporators
Product Type Electroporation accessory
Voltage 200 V
Description Genetrodes, 1 mm L-Shape (GOLD TIP)
Electrode Shape Bent L-shaped
Includes Genetrode only
Length (English) Electrode 0.039 in.
Length (Metric) Electrode 1 mm
Life Span ∼1500+ Pulses
Pulse Number Range 1 sec. to 99 msec.
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 1 Ea.
Size (English) Gap 0.03 to 0.39 in.
Size (Metric) Gap 1 to 10 mm
Tip Type Gold Tip
Sterility Non-sterile
Connection Type Banana Plug to Micrograbber
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.