Recombinant Protein;200UG;Biotinylated Human BTLA (31-134), Fc,Avitag (BTA-H82F8) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession # Q7Z6A9).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 40.5 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 10 g/mL (100 L/well) can bind Biotinylated Human BTLA (31-134), Fc,Avitag (Cat. No. BTA-H82F8) with a linear range of 0.3-5 g/mL (QC tested).