missing translation for 'onlineSavingsMsg'
Learn More

BTEX/MTBE in Soil - PT, MilliporeSigma™ Supelco™

Catalog No. 111001679
Change view
Click to view available options
Quantity:
30 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001679 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001679 Supplier Merck Emd Millipore Supplier No. SPE02530G
Only null left

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 67-56-1
Shipping Condition Wet Ice
Type Proficiency Testing Material
Quantity 30 g