New England Biolab Ltd Bsu DNA Polymerase, Large Fragment 1000 units
Numéro de catalogue. 504368
Bsu DNA Polymerase I, Large Fragment retains the 5' to 3' polymerase activity of the Bacillus subtilis DNA polymerase I, but lacks the 5' to 3' exonuclease domain. This large fragment naturally lacks 3' to 5' exonuclease activity.
Numéro de catalogue.
504368
Fournisseur
New England Biolab LtdCode fournisseur
M0330L
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Bsu DNA Polymerase I, Large Fragment retains the 5' to 3' polymerase activity of the Bacillus subtilis DNA polymerase I, but lacks the 5' to 3' exonuclease domain. This large fragment naturally lacks 3' to 5' exonuclease activity.